Digalliumtrioksidi ≥99.999% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific

11151.06 11151.14 11151.22 11151.36
11151.06EA 188 EUR
11151.06 11151.14 11151.22 11151.36
Digalliumtrioksidi ≥99.999% (metals basis), Puratronic®
Digalliumtrioksidi

Purity: 99.999% CAS No.: 12024-21-4 Molecular Formula: Ga2O3 Formula Weight: 187.44 Physical Form: -50 Mesh Powder Density: 6.44 Refractive Index: 1.92 Odour: Odourless Melting Point: 1900° Merck Reference: 14,4346 MDL No.: MFCD00011020

Kaava: Ga₂O₃
Sulamispiste: 1900 °C
Tiheys: 6,44
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011020
CAS-numero: 12024-21-4
EINECS: 234-691-7
Merck Index: 14,4346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR