Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)

Toimittaja: Thermo Fisher Scientific

11144.06 11144.14 11144.22
11144.06EA 306 EUR
11144.06 11144.14 11144.22
Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)
Germanium

Purity: 99.9999+% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odour: Odourless MDL No.: MFCD00085310

Kaava: Ge
MW: 72.61 g/mol
Kiehumispiste: 2830
Sulamispiste: 937.2
Tiheys: 5.323 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00085310
CAS-numero: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR