VWR®, Silicone Lab Mat
Toimittaja: VWR Collection
111-9200EA 70.4 EUR
111-9200
VWR®, Silicone Lab Mat
Suojapaperit ja pintasuojukset Pöydänsuojukset
The reusable, environmentally friendly lab mat is an economical solution to help keep bench tops clean and safe from stains, spills and wear. The Lab Mat is made from a durable FDA-approved silicone material that creates a stain resistant, washable working surface. It is chemically inert and does not react with most chemicals.
  • Protect bench tops from hot items up to 200 °C
  • Large anti-skid and noise-dampening working surface
  • Lip design around the outer edge of the mat helps contain spillage
  • Can be cleaned with standard disinfectants in a sink or dishwasher,