Naamiomalliset suojalasit, uvex ultrasonic
Toimittaja: UVEX
9302.285 9302.600 9302.255 9302.245 9302.281 9302.286 9302.275
111-8009EA 41.9 EUR
111-8009 111-1911 111-8222 111-8008 111-8064 UVEX9302.286 111-1673
Naamiomalliset suojalasit, uvex ultrasonic
Suojalasit, naamiomalliset
These full view goggles fit individual facial features well, due to their combination of hard and soft materials, ensuring a high level of comfort for the wearer.
  • Optimised internal volume and extensive side protection, even for spectacle wearers
  • Prescription insert possible
  • Wide panoramic field of view
  • Ventilated for a pleasant and irritation-free interior environment
Paino: 80 g.
* heikompi ylä- ja alatuuletus ** pääpanta on kumia.
** Model has rubber headband.
Sertifikaatit: EN 166, EN 170; 1B.