Uvex Ultravision Goggles
Toimittaja: UVEX
9301.105 9301.317 9301.714 9301.906 9301.716 9301.318 9301.555 9301.626 9301.116
UVEX9301.105EA 41.9 EUR
UVEX9301.105 111-8049 111-8051 UVEX9301.906 UVEX9301.716 UVEX9301.318 UVEX9301.555 111-1713 UVEX9301.116
Uvex Ultravision Goggles
Suojalasit, naamiomalliset
Goggles with panoramic lens shape allow 180° uninterrupted lateral vision. They are anatomically shaped and fit comfortably over most prescription spectacles.
  • Integrated, indirect ventilation allows air movement
  • Frosted top to reduce glare
  • Specially designed nose bridge, works well with respirator
  • Wide, high-quality, easily adjustable headband
  • Additional lightweight face shield with integrated ventilation to clip onto goggles
  • Fits over breathing filter, safety helmets and ear muffs
Weight: 126 g
EN 166, EN 170
Sertifikaatit: EN 166, EN 170.