Face shields
Toimittaja: UVEX
9707.014 9708.514
111-1577EA 78.7 EUR
111-1577 UVEX9708.514
Face shields
Kasvosuojaimet
Robust visor with integrated forehead cover, clear cellulose acetate lens and additional protective cap made from impact-resistant PS.
  • Twist ratchet enables simple and reliable adjustment to any head size
  • Flip-up mechanism allows the visor to be set in any position
  • Easy lens replacement
Sertifikaatit: EN 166