Puolimaski, hiukkasia suodattava, Comfort range
Toimittaja: HONEYWELL SAFETY
1007222 1007221
111-3533EA 43.7 EUR
111-3533 111-1269
Puolimaski, hiukkasia suodattava, Comfort range
Hengityssuojaimet Kertakäyttöiset hengityssuojaimet Hengityssuojaimet
These half masks protect against dusts, mists and fumes. Latex-, silicone-, PVC-free. Ideal for use in textile, construction, iron and steel industries.
  • Rigid shell for a shape maintained even in moistened environments
  • Pre-formed and coloured nose bridge for a better fit and an instant identification of the protection level of the respirator
  • Large internal volume for a better comfort and easier communication
  • Twin elastic straps, ultra-flexible and strong, stapled outside the filtering area to avoid any risk of leakages
  • Lightweight respirator (10 g) for a