Goggles, Honeywell LG
Toimittaja: HONEYWELL SAFETY
1005507 1005509 1005504 1008112
111-1231EA 5.1 EUR
111-1231 111-1232 DALL1005504 DALL1008112
Goggles, Honeywell LG
Suojalasit, naamiomalliset
The impact resistant polycarbonate lens offers an excellent protection against flying particles, coarse dusts, liquid droplets, mists and sprays.
  • Lightweight design and large field of vision
  • Soft seal and easily adjustable headband: Fits a wide variety of workers
  • With or without ventilation
The acetate, anti-fog lens and neoprene headband offers superior chemical resistance allowing the use in a broad range of chemical industries.
Weight: 75 to 79 g
Sertifikaatit: EN 166