Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

11042.04 11042.09
11042.04EA 454 EUR
11042.04 11042.09
Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9% (metals basis)
Ammonium hexachloropalladate(IV)

Purity: 99.9% CAS No.: 19168-23-1 Molecular Formula: (NH4)2PdCl6 Formula Weight: 355.20 Physical Form: Crystalline Density: 2.42 Appearance: Red-orange Odour: Odourless UN No.: 3077 MDL No.: MFCD00015959

Kaava: H₈Cl₆N₂Pd
Tiheys: 2,42
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00015959
CAS-numero: 19168-23-1
EINECS: 242-854-9
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR