Rhodium(III) iodide (≥20,8% Rh) ≥99.9% (metals basis)

Toimittaja: Thermo Fisher Scientific
Varoitus

11029.01 11029.03 11029.06
11029.01EA 330 EUR
11029.01 11029.03 11029.06
Rhodium(III) iodide (≥20,8% Rh) ≥99.9% (metals basis)
Rhodium(III) iodide

Purity: 99.9% CAS No.: 15492-38-3 Molecular Formula: RhI3 Formula Weight: 483.62 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Black Odour: Odourless MDL No.: MFCD00016311

Kaava: I₃Rh
Kiehumispiste: 100 °C (1013 hPa)
Tiheys: 6,4 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016311
CAS-numero: 15492-38-3
EINECS: 239-521-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR