Natriumfluoridi, Optical Grade

Toimittaja: Thermo Fisher Scientific
Vaara

11023.18
11023.18EA 809 EUR
11023.18
Natriumfluoridi, Optical Grade
Natriumfluoridi

CAS No.: 7681-49-4 Molecular Formula: NaF Formula Weight: 41.99 Physical Form: Broken Crystals Density: 2.78 Refractive Index: 1.336 Melting Point: 993° Boiling Point: 1704° Merck Reference: 14,8618 UN No.: 1690 MDL No.: MFCD00003524

Kaava: NaF
Kiehumispiste: 1704 °C
Sulamispiste: 993 °C
Tiheys: 2,78
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003524
CAS-numero: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 14,8618
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR