VWR® Ready to Use Media Plates (90 mm), Irradiated
Toimittaja: VWR Chemicals
111114ZIEA 34.9 EUR
111114ZI 131114ZI 131114PY 131114RY 111114TI 131114TI 110802ZI 130802ZI 110884ZI 130884ZI 110774ZI 110070PY 110884CY 112750ZI 130884CY 110884TI 110802ZIMP 110884TIMP 110774ZIMP 111114TIMP 111114ZIMP 112230CAMP 131114TIMP 131114ZIMP 131114PYMP 281112ZI 280882TI 280882ZI 281112TI 281112TI30 499270OO 131114XI 285712TI
VWR® Ready to Use Media Plates (90 mm), Irradiated
Elatusaineet mikrobiologiaan Valmisteltu viljelyaine
Saatavissa standardimaljoissa (halkaisija 90 mm). Neutralointiaineena on käytetty TLHTh:a (Tween, lesitiini, histidiini, tiosulfaatti). Pussit saa kaksin- tai kolminkertaisesti pakattuina. Toisessa pakkauksen pusseista on säteilytystiedot.
  • Triple-wrapping ensures that the package itself doesn't contaminate the environment, as the first wrapper is removed just before entering a clean area
  • Available with certificate of irradiation/dosimetry
  • VHP resistance studies available for all triple-wrapped plates
Storage: 15 to 25 °C for plates 280882TI, 280882ZI, 281112TI and 281112ZI; 2 to 14 °C for all the other plates.
Pakkaus: The plates are packed 10 each in a plastic shrink-wrapped bag. Box contains 2 bags with a total of 20 or 6×10 plates.