Affinity chromatography columns, His GraviTrap™
Toimittaja: Cytiva
11-0033-99
11-0033-99EA 323 EUR
11-0033-99
Affinity chromatography columns, His GraviTrap™
Kromatografiakolonnit
His GraviTrap is a prepacked, single-use column for purification of histidine-tagged proteins by immobilized metal affinity chromatography (IMAC). Column gives fast and simple gravity-flow purifications without any need for a purification.
  • For fast and simple purification of histidine-tagged proteins
  • Convenient, prepacked gravity-flow column
  • Allows direct purification of both clarified and unclarified cell lysates
  • High binding capacity; approximately 40 mg of pure protein per column
  • Compatible with a wide range of reducing agents, detergents, denaturants