Kaliumkarbonaatti-1,5 H2O 98.5-101.0% ACS

Toimittaja: Thermo Fisher Scientific
Varoitus

10981.22 10981.36
10981.22EA 89.5 EUR
10981.22 10981.36
Kaliumkarbonaatti-1,5 H2O 98.5-101.0% ACS
Kaliumkarbonaatti-1,5 H2O

Purity: 98.5-101.0% CAS No.: 6381-79-9 Molecular Formula: K2CO3.1.5H2O Formula Weight: 165.23 (138.21anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Solubility: Soluble in water. Practically insoluble in alcohol Merck Reference: 14,7670 MDL No.: MFCD00149925


Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent

Kaava: CK₂O₃·1,5H₂O
Sulamispiste: 891 °C
Tiheys: 2,29 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149925
CAS-numero: 6381-79-9
EINECS: 209-529-3
Merck Index: 14,7670

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR