Palladium ≥99.99% (metals basis), wire, Premion®, Ø 0.1 mm

Toimittaja: Thermo Fisher Scientific

10963.G5 10963.G9
10963.G5EA 366 EUR
10963.G5 10963.G9
Palladium ≥99.99% (metals basis), wire, Premion®, Ø 0.1 mm
Palladium

Purity: 99.99% CAS No.: 7440-05-3 Physical Form: ≈0.09g/m Odour: Odourless MDL No.: MFCD00011167

Kaava: Pd
MW: 106.420 g/mol
Tiheys: 12.02 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011167
CAS-numero: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR