Terbium(III,IV)oksidi ≥99.998% (REO, rare earth oxide basis), REacton®

Toimittaja: Thermo Fisher Scientific

10917.03 10917.06 10917.14 10917.22
10917.03EA 194 EUR
10917.03 10917.06 10917.14 10917.22
Terbium(III,IV)oksidi ≥99.998% (REO, rare earth oxide basis), REacton®
Terbium(III,IV)oksidi

Purity: 99.998% CAS No.: 12037-01-3 Molecular Formula: Tb4O7 Formula Weight: 747.69 Physical Form: Powder Odour: Odourless Merck Reference: 14,9157 MDL No.: MFCD00083151

Kaava: O₇Tb₄
Sulamispiste: 2340 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00083151
CAS-numero: 12037-01-3
EINECS: 234-856-3
Merck Index: 14,9157

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR