Ammonium bis(oxalato)oxotitanate(IV) hydrate ≥99.998% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Varoitus

10895.09 10895.18
10895.09EA 200 EUR
10895.09 10895.18
Ammonium bis(oxalato)oxotitanate(IV) hydrate ≥99.998% (metals basis), Puratronic®
Ammonium bis(oxalato)oxotitanate(IV) hydrate

Purity: 99.998% CAS No.: 10580-03-7 Molecular Formula: (NH4)2TiO(C2O4)2.xH2O (x≈1) Formula Weight: 276.01(anhy) Physical Form: Crystalline powder Odour: Odourless UN No.: UN3288 MDL No.: MFCD00149620

Kaava: (NH₄)₂TiO(C₂O₄)₂·xH₂O (x≈1)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149620
CAS-numero: 10580-03-7
EINECS: 234-174-6
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR