Rubidiumkloridi 99.975% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Rubidiumchloride

10851.09 10851.18
10851.09EA 446 EUR
10851.09 10851.18
Rubidiumkloridi 99.975% (metals basis), Puratronic®
Rubidiumkloridi

Purity: 99.975% CAS No.: 7791-11-9 Molecular Formula: RbCl Formula Weight: 120.92 Physical Form: Crystalline Aggregates Density: 2.800 Refractive Index: 1.493 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 718° Boiling Point: 1390° Merck Reference: 14,8284 MDL No.: MFCD00011187

Kaava: RbCl
MW: 120.92 g/mol
Kiehumispiste: 1390 °C
Sulamispiste: 718 °C
Tiheys: 2.800
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011187
CAS-numero: 7791-11-9
EINECS: 232-240-9
Merck Index: 14,8284

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR