Nickel sulphate hydrate ≥99.9985% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Vaara

10820.14 10820.22
10820.14EA 266 EUR
10820.14 10820.22
Nickel sulphate hydrate ≥99.9985% (metals basis), Puratronic®
Nickel sulphate hydrate

Purity: 99.9985% CAS No.: 15244-37-8 Molecular Formula: NiO4S.xH2O (x≈6) Formula Weight: 154.77(anhy) Physical Form: Crystalline powder Density: 2.070 Appearance: Green Odour: Odourless pH: 4.3-4.7 Merck Reference: 14,6517 UN No.: 3288 MDL No.: MFCD00011146

Kaava: NiO₄S·H₂O
Tiheys: 2,070
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011146
CAS-numero: 15244-37-8
EINECS: 232-104-9
UN: 3288
ADR: 6.1,III
Merck Index: 14,6517

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR