Nikkelidinitraattihexahydraatii ≥99.9985% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Vaara

10816.06 10816.18 10816.30
10816.06EA 158 EUR
10816.06 10816.18 10816.30
Nikkelidinitraattihexahydraatii ≥99.9985% (metals basis), Puratronic®
Nikkelidinitraattihexahydraatii

Purity: 99.9985% CAS No.: 13478-00-7 Molecular Formula: N2NiO6.6H2O Formula Weight: 290.80 (182.70anhy) Physical Form: Crystalline Aggregates Density: 2.05 Appearance: Green Odour: Odourless Melting Point: 56.7° Boiling Point: 137° Merck Reference: 14,6513 UN No.: 2725 MDL No.: MFCD00149805

Kaava: Ni(NO₃)₂·6H₂O
Kiehumispiste: 137 °C
Sulamispiste: 56,7 °C
Tiheys: 2,05
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149805
CAS-numero: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1,III
Merck Index: 14,6513

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR