Magnesium nitrate hydrate ≥99.999% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Vaara

10799.14 10799.22 10799.36 10799.06
10799.14EA 233 EUR
10799.14 10799.22 10799.36 10799.06
Magnesium nitrate hydrate ≥99.999% (metals basis), Puratronic®
Magnesium nitrate hydrate

It is used in printing, chemical, agriculture and ceramics industries. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.


Purity: 99.999% CAS No.: 10377-60-3 Molecular Formula: Mg(NO3)2.xH2O Formula Weight: 148.32(anhy) Physical Form: Crystalline Aggregates Density: 1.46 Melting Point: 89° Boiling Point: 330° dec. Merck Reference: 14,5674 UN No.: 1474 MDL No.: MFCD00011103

Kaava: Mg(NO₃)₂·xH₂O
MW: 148.32 g/mol (anhy)
Kiehumispiste: 330 °C (haj. lämpötila)
Tiheys: 1.46
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011103
CAS-numero: 10377-60-3
EINECS: 233-826-7
UN: 1474
ADR: 5.1 III
Merck Index: 14,5674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR