tri-Litiumortofosfaatti ≥99.99% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Varoitus

10743.14 10743.22
10743.14EA 435 EUR
10743.14 10743.22
tri-Litiumortofosfaatti ≥99.99% (metals basis), Puratronic®
tri-Litiumortofosfaatti

Purity: 99.99% CAS No.: 10377-52-3 Molecular Formula: Li3PO4 Formula Weight: 115.79 Physical Form: Powder Density: 2.54 Appearance: White Odour: Odourless Melting Point: 837° MDL No.: MFCD00016187

Kaava: Li₃O₄P
Sulamispiste: 837 °C
Tiheys: 2,54
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016187
CAS-numero: 10377-52-3
EINECS: 233-823-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR