Diammonium hexachloroiridate (41% Ir) ≥99.994% (metals basis) Ir ≥ 41%, Premion®

Toimittaja: Thermo Fisher Scientific
Varoitus

10712.03 10712.06 10712.77
10712.03EA 793 EUR
10712.03 10712.06 10712.77
Diammonium hexachloroiridate (41% Ir) ≥99.994% (metals basis) Ir ≥ 41%, Premion®
Diammonium hexachloroiridate

Purity: 99.994% CAS No.: 16940-92-4 Molecular Formula: (NH4)2IrCl6 Formula Weight: 441.02 Physical Form: Crystalline powder Density: 2.86 Appearance: Red-black Melting Point: dec. MDL No.: MFCD00010881

Kaava: H₈Cl₆IrN₂
Sulamispiste: Dec.
Tiheys: 2,86
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010881
CAS-numero: 16940-92-4
EINECS: 241-007-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR