Indium(III)oksidi ≥99.997% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific

Synonyms: Indium trioxide, Diindium trioxide

10709.09 10709.18
10709.09EA 299 EUR
10709.09 10709.18
Indium(III)oksidi ≥99.997% (metals basis), Puratronic®
Indium(III)oksidi

Purity: 99.997% CAS No.: 1312-43-2 Molecular Formula: In2O3 Formula Weight: 277.64 Physical Form: Powder Density: 7.18 Melting Point: 850° volat. Merck Reference: 14,4952 MDL No.: MFCD00011060

Kaava: In₂O₃
MW: 277.64 g/mol
Sulamispiste: 850 °C
Tiheys: 7.18
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011060
CAS-numero: 1312-43-2
EINECS: 215-193-9
Merck Index: 14,4952

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR