Kromi(III)oksidi ≥99.97% (metals basis), jauhe, Puratronic® -22 Mesh

Toimittaja: Thermo Fisher Scientific
Varoitus

10688.06 10688.14 10688.22 10688.36
10688.06EA 147 EUR
10688.06 10688.14 10688.22 10688.36
Kromi(III)oksidi ≥99.97% (metals basis), jauhe, Puratronic® -22 Mesh
Kromi(III)oksidi

Purity: 99.97% CAS No.: 1308-38-9 Molecular Formula: Cr2O3 Formula Weight: 151.99 Physical Form: -22 Mesh Powder Density: 5.22 Refractive Index: 2.551 Appearance: Pale to dark green Odour: Odourless Melting Point: 2435° Boiling Point: 3000° Merck Reference: 14,2234 MDL No.: MFCD00010949

Kaava: Cr₂O₃
Kiehumispiste: 3000 °C
Sulamispiste: 2435 °C
Tiheys: 5,22
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010949
CAS-numero: 1308-38-9
EINECS: 215-160-9
Merck Index: 14,2234

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR