Kalsiumfluoridi ≥99.985% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Varoitus

10681.09 10681.18
10681.09EA 465 EUR
10681.09 10681.18
Kalsiumfluoridi ≥99.985% (metals basis), Puratronic®
Kalsiumfluoridi

Purity: 99.985% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: Powder Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 1403° Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

Kaava: CaF₂
Kiehumispiste: ∼2500 °C
Sulamispiste: 1403 °C
Tiheys: 3,18
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010907
CAS-numero: 7789-75-5
EINECS: 232-188-7
Merck Index: 14,1667

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR