Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Varoitus

10680.04 10680.09 10680.18 10680.30
10680.04EA 116 EUR
10680.04 10680.09 10680.18 10680.30
Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®
Calcium chloride hydrate

Purity: 99.9965% CAS No.: 7774-34-7 Molecular Formula: CaCl2.xH2O (x≈4-6) Formula Weight: 110.99(anhy) Physical Form: Crystalline Aggregates Density: 1.71 Merck Reference: 14,1659 MDL No.: MFCD00149616

Kaava: CaCl₂·H₂O
Sulamispiste: 29,54 °C
Tiheys: 1,71
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149613
CAS-numero: 22691-02-7
EINECS: 607-129-7
Merck Index: 14,1659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR