Renium 99,997 Metals basis, powder, Puratronic® -22 mesh

Toimittaja: Thermo Fisher Scientific
Varoitus

10673.03 10673.06
10673.03EA 252 EUR
10673.03 10673.06
Renium 99,997 Metals basis, powder, Puratronic® -22 mesh
Renium

Purity: 99.997% CAS No.: 7440-15-5 UN No.: 3089 MDL No.: MFCD00011195

Kaava: Re
MW: 186.21 g/mol
Kiehumispiste: 5630
Sulamispiste: 3180
Tiheys: 21.02
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011195
CAS-numero: 7440-15-5
EINECS: 231-124-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,08343

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR