Zirconium ≥99.2% (metals basis excluding Hf), plate (max. 4.5% Hf), Thickness: 3.175 mm (0.125 in)

Toimittaja: Thermo Fisher Scientific

10593.GHEA 779 EUR
10593.GH
Zirconium ≥99.2% (metals basis excluding Hf), plate (max. 4.5% Hf), Thickness: 3.175 mm (0.125 in)
Zirconium

Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈206.6g/100x100mm Odour: Odourless MDL No.: MFCD00011303

Kaava: Zr
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011303
CAS-numero: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR