Rhodium(II) acetate dimer ≥99.99% (metals basis) Rh ≥ 46.2%, Premion®

Toimittaja: Thermo Fisher Scientific

10560.01 10560.03 10560.06
10560.01EA 705 EUR
10560.01 10560.03 10560.06
Rhodium(II) acetate dimer ≥99.99% (metals basis) Rh ≥ 46.2%, Premion®
Rhodium(II) acetate dimer

Purity: 99.99% CAS No.: 15956-28-2 Molecular Formula: C8H12O8Rh2 Formula Weight: 441.99 Physical Form: Powder Appearance: Green Odour: Odourless Melting Point: 205° Solubility: Slightly soluble in ethanol MDL No.: MFCD00003538


Cyclopropanation of alkenes

Kaava: C₈H₁₂O₈Rh₂
Sulamispiste: 205 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003538
CAS-numero: 15956-28-2
EINECS: 240-084-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR