Potassium tetracyanoplatinate(II) trihydrate (≥44,9% Pt) ≥99.9% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

10536.03 10536.06
10536.03EA 411 EUR
10536.03 10536.06
Potassium tetracyanoplatinate(II) trihydrate (≥44,9% Pt) ≥99.9% (metals basis)
Potassium tetracyanoplatinate(II) trihydrate

Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.


Soluble in hot water

Kaava: C₄K₂N₄Pt·3H₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149921
CAS-numero: 38245-27-1
EINECS: 209-236-0
UN: 1588
ADR: 6.1,III
Merck Index: 14,7685

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR