cis-Dichlorobis(triphenylphosphite)platinum(II) (≥24,2% Pt)

Toimittaja: Thermo Fisher Scientific
Varoitus

10492.03 10492.MD
10492.03EA 378 EUR
10492.03 10492.MD
cis-Dichlorobis(triphenylphosphite)platinum(II) (≥24,2% Pt)
cis-Dichlorobis(triphenylphosphite)platinum(II)

CAS No.: 15604-36-1 Molecular Formula: PtCl2[P(C6H5)3]2 Formula Weight: 790.58 Physical Form: Crystalline Sensitivity: Hygroscopic Melting Point: 310° dec. MDL No.: MFCD00010825

Kaava: [(C₆H₅)₃P]₂PtCl₂
Sulamispiste: 310 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010825
CAS-numero: 15604-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR