Alumiinioksidi, γ-α-faasi ≥99.98% (metals basis)

Toimittaja: Thermo Fisher Scientific

10459.14 10459.22 10459.36
10459.14EA 107 EUR
10459.14 10459.22 10459.36
Alumiinioksidi, γ-α-faasi ≥99.98% (metals basis)
Alumiinioksidi, γ-α-faasi

Purity: 99.98% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 0.01-0.02 Micron Powder, S.A. 100m2/g Density: 3.965 Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424

Kaava: Al₂O₃
Kiehumispiste: 2980 °C
Sulamispiste: 2045 °C
Tiheys: 3,965
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003424
CAS-numero: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR