Zirconium ≥99% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 6.2 mm (0.24 in)

Toimittaja: Thermo Fisher Scientific

10443.BSEA 128 EUR
10443.BS 10443.BW
Zirconium ≥99% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 6.2 mm (0.24 in)
Zirconium

Purity: 99% CAS No.: 7440-67-7 Physical Form: ≈1.96g/cm, Temper: soft Odour: Odourless MDL No.: MFCD00011303

Kaava: Zr
MW: 91.22 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011303
CAS-numero: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR