Volframi 99,9 Metals basis, powder APS 12 µm

Toimittaja: Thermo Fisher Scientific
Varoitus

10401.22
10401.22EA 156 EUR
10401.22
Volframi 99,9 Metals basis, powder APS 12 µm
Volframi

Purity: 99.9% CAS No.: 7440-33-7 Physical Form: Powder Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461

Kaava: W
MW: 183.84 g/mol
Kiehumispiste: 5660
Sulamispiste: 3410
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011461
CAS-numero: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1 III
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR