Platina, lanka for ISA type R or S standard grade thermocouple, Ø 0.127 mm (0.005 in)

Toimittaja: Thermo Fisher Scientific

10289.G1 10289.G5 10289.G9
10289.G1EA 371 EUR
10289.G1 10289.G5 10289.G9
Platina, lanka for ISA type R or S standard grade thermocouple, Ø 0.127 mm (0.005 in)
Platina

CAS No.: 7440-06-4 Physical Form: ≈0.27g/m Odour: Odourless MDL No.: MFCD00011179

Kaava: Pt
Kiehumispiste: 3827 °C (1013 hPa)
Sulamispiste: 1769 °C
Tiheys: 21,447 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011179
CAS-numero: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR