Magnesium ≥99.8%, jauhe -325 mesh

Toimittaja: Thermo Fisher Scientific
Vaara

10233.09 10233.22 10233.36 10233.57
10233.09EA 167 EUR
10233.09 10233.22 10233.36 10233.57
Magnesium ≥99.8%, jauhe -325 mesh
Magnesium

Purity: 99.8% CAS No.: 7439-95-4 Appearance: Silver-grey Odour: Odourless UN No.: 1418 MDL No.: MFCD00085308

Kaava: Mg
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00085308
CAS-numero: 7439-95-4
EINECS: 231-104-6
UN: 1418
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR