Rauta ≥99.97% (metals basis), pieces, irregularly shaped ≤ 25 mm (1.0 in)

Toimittaja: Thermo Fisher Scientific

10213.30 10213.A1 10213.A7 10213.PG_25KG
10213.30EA 116 EUR
10213.30 10213.A1 10213.A7 10213.PG_25KG
Rauta ≥99.97% (metals basis), pieces, irregularly shaped ≤ 25 mm (1.0 in)
Rauta

Purity: 99.97+% CAS No.: 7439-89-6 Physical Form: 90% 2.5-25mm (0.1-1.0in) Appearance: Gray MDL No.: MFCD00010999

Kaava: Fe
Kiehumispiste: 3000 °C (1013 hPa)
Sulamispiste: 1535 °C
Tiheys: 7,86 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010999
CAS-numero: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR