Kromi ≥99% (metals basis), irregular pieces

Toimittaja: Thermo Fisher Scientific

10150.22 10150.36
10150.22EA 182 EUR
10150.22 10150.36
Kromi ≥99% (metals basis), irregular pieces
Kromi

Purity: 99% CAS No.: 7440-47-3 Physical Form: Broken plate Odour: Odourless MDL No.: MFCD00010944

Kaava: Cr
Kiehumispiste: 2469 °C (1013 hPa)
Sulamispiste: 1840 °C
Tiheys: 7,2 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010944
CAS-numero: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR