Formalin 10%, Q Path®, buffered

Toimittaja: VWR Q-Path Chemicals
Vaara

Synonyms: Formaldehydi

FOR0020AF59001EA 89.4 EUR
FOR0020AF59001 FOR0150AF59001 FOR0060AF59001 10099464. FOR0070AF59001 11699455. 10099465. 11699404. 11699408. FOR010LAF59001 VWRQ10.02010WH0001 VWRQ10.07040WH0002 VWRQ10.15060RD0001 VWRQ10.1000.0002 VWRQ10.5000.0001 VWRQ10.10000.0001 VWRQ10.06030WH0001 10.300150WH01
Formalin 10%, Q Path®, buffered
Formalin

Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.


Formaldehyde 4% is approximately equal to Formalin 10%.

Kaava: CH₂O
MW: 30.03 g/mol
Tiheys: 1.013 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003274
CAS-numero: 50-00-0
EINECS: 200-001-8
IVDLogo

Order Now

Testitulosten erittely

Appearance Colourless liquid
Test formaldehyde Passes test
Density (20/4) 1,012 - 1,014
pH (20°C) 6,8 - 7,2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR