Antimoni ≥99.5% (metals basis), jauhe -200 mesh

Toimittaja: Thermo Fisher Scientific
Vaara

10099.09 10099.22 10099.36 10099.A3 10099.PH
10099.09EA 127 EUR
10099.09 10099.22 10099.36 10099.A3 10099.PH
Antimoni ≥99.5% (metals basis), jauhe -200 mesh
Antimoni

Purity: 99.5% CAS No.: 7440-36-0 Appearance: Gray Odour: Odourless UN No.: 2871 MDL No.: MFCD00134030

Kaava: Sb
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134030
CAS-numero: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR