Bis(benzonitrile)dichloroplatinum(II) (≥40% Pt)

Toimittaja: Thermo Fisher Scientific
Vaara

10004.02 10004.03 10004.06
10004.02EA 273 EUR
10004.02 10004.03 10004.06
Bis(benzonitrile)dichloroplatinum(II) (≥40% Pt)

CAS No.: 14873-63-3 Molecular Formula: PtCl2[(C6H5)(CN)]2 Formula Weight: 472.25 Physical Form: Powder Solubility: Slightly soluble in acetone, chloroform UN No.: 3439 MDL No.: MFCD00013125

Kaava: C₁₄H₁₀Cl₂N₂Pt
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013125
CAS-numero: 14873-63-3
EINECS: 238-943-7
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR