Supelco® TLC Explorer Documentation System
Toimittaja: Merck
1.52610.0001 1.52611.0001
1.52610.0001EA 10040 EUR
1.52610.0001 1.52611.0001
Supelco® TLC Explorer Documentation System
Fotodokumentointijärjestelmät
Superior instrumental solution for reliable TLC plate analysis. It can analyse one or multiple plates up to an area of 20×20 cm. In addition, the device uses three LED light sources – white light (VIS), UV-A (366 nm), and UV-C (254 nm) to illuminate the plates from above in direct-light mode. It allows a quick visual check or a software-based analysis and quantification.
  • Small, portable with off -grid functionality
  • Safe and secure handling
  • Sustainable – long-lasting LED light sources with low energy usage
  • Flexible software and connection
  • Reliable analysis