HPLC-kolonni, Chromolith®
Toimittaja: Merck
1.02129.0001 1.51450.0001 1.51463.0001 1.51466.0001 1.51451.0001 1.51452.0001 1.51465.0001 1.51468.0001 1.52001.0001 1.52007.0001 1.52003.0001 1.52004.0001 1.52005.0001 1.52006.0001 1.52002.0001 1.52008.0001 1.52009.0001 1.52014.0001 1.52013.0001 1.52035.0001 1.52063.0001 1.52062.0001
1.02129.0001EA 1050 EUR
1.02129.0001 1.51450.0001 1.51463.0001 1.51466.0001 1.51451.0001 1.51452.0001 1.51465.0001 1.51468.0001 1.52001.0001 1.52007.0001 1.52003.0001 1.52004.0001 1.52005.0001 1.52006.0001 1.52002.0001 1.52008.0001 1.52009.0001 1.52014.0001 1.52013.0001 1.52035.0001 1.52063.0001 1.52062.0001
HPLC-kolonni, Chromolith®
Kromatografiakolonnit
Monolithic silica technology: A new age in chromatography. Chromolith® columns are based on silica so existing methods can be easily transferred with only minimal investment in new method development work. 'Sol-Gel' technology allows highly porous monolithic rods of silica to be formed. These feature a bimodal pore structure provides a unique combination of macropores and mesopores.
  • Due to their extremely high porosity, very high flow rates can be used with low pressures
  • Reduces separation time for high speed analysis
  • Long column and HPLC system lifetime