HPLC-kolonni, Hibar®, LiChrospher® Si 60
Toimittaja: Merck
1.00423.7104 1.00424.7104 1.50012.7104 1.50013.7104 1.50182.7104 1.50183.7104 1.00424.7109 1.50183.7106 1.51196.7106 1.51196.7109 1.50182.7106 1.50013.7109
1.00423.7104EA 1120 EUR
1.00423.7104 1.00424.7104 1.50012.7104 1.50013.7104 1.50182.7104 1.50183.7104 1.00424.7109 1.50183.7106 1.51196.7106 1.51196.7109 1.50182.7106 1.50013.7109
HPLC-kolonni, Hibar®, LiChrospher® Si 60
Kromatografiakolonnit
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.