HPLC-kolonni, LiChroCART®, Superspher® 100 RP-18e
Toimittaja: Merck
1.50171.7138 1.50173.7138 1.50178.7138 1.50179.7138 1.50201.7138 1.50228.7138 1.50229.7138 1.50233.7138 1.50234.7138 1.50236.7138 1.51442.7138 1.51443.7138 1.51444.7138 1.51448.7138
1.50171.7138EA 683 EUR
1.50171.7138 1.50173.7138 1.50178.7138 1.50179.7138 1.50201.7138 1.50228.7138 1.50229.7138 1.50233.7138 1.50234.7138 1.50236.7138 1.51442.7138 1.51443.7138 1.51444.7138 1.51448.7138
HPLC-kolonni, LiChroCART®, Superspher® 100 RP-18e
Kromatografiakolonnit
Stainless steel LiChroCART® cartridges are available with various types of Superspher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are asMore Product Information