HPLC-kolonni, LiChroCART®, LiChrospher® 100 DIOL
Toimittaja: Merck
1.50201.7075 1.50234.7075 1.50236.7075 1.51432.7075 1.50177.7075 1.51443.7075 1.51444.7075 1.51445.7075 1.51431.7075 1.50179.7075 1.50228.7075 1.50229.7075 1.50233.7075 1.50173.7075 1.51442.7075 1.50171.7075 1.51448.7075 1.51449.7075 1.50178.7075
1.50201.7075EA 0 EUR
1.50201.7075 1.50234.7075 1.50236.7075 1.51432.7075 1.50177.7075 1.51443.7075 1.51444.7075 1.51445.7075 1.51431.7075 1.50179.7075 1.50228.7075 1.50229.7075 1.50233.7075 1.50173.7075 1.51442.7075 1.50171.7075 1.51448.7075 1.51449.7075 1.50178.7075
HPLC-kolonni, LiChroCART®, LiChrospher® 100 DIOL
Kromatografiakolonnit
Stainless steel LiChroCART® cartridges are available with LiChrospher® sorbents. Ideal for separation of compounds under basic eluents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separateMore Product Information