HPLC-kolonni, Hibar®, LiChrospher® 100 CN
Toimittaja: Merck
1.00423.7070 1.00424.7070 1.50009.7070 1.50012.7070 1.50013.7070 1.50182.7070 1.50183.7070 1.00424.7071 1.50182.7071 1.50009.7071 1.50183.7071 1.50013.7071
1.00423.7070EA 1120 EUR
1.00423.7070 1.00424.7070 1.50009.7070 1.50012.7070 1.50013.7070 1.50182.7070 1.50183.7070 1.00424.7071 1.50182.7071 1.50009.7071 1.50183.7071 1.50013.7071
HPLC-kolonni, Hibar®, LiChrospher® 100 CN
Kromatografiakolonnit
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.