HPLC-kolonni, Hibar®, with Purospher® Sorbents
Toimittaja: Merck
1.00423.7130 1.00424.7130 1.50009.7127 1.50009.7130 1.50012.7130 1.50013.7127 1.50013.7130 1.50013.7184 1.50182.7127 1.50182.7130 1.50183.7127 1.50183.7130 1.50009.7184 1.50183.7185 1.50182.7174 1.00423.7184 1.50009.7220 1.00424.7184 1.50183.7177 1.00424.7175 1.00424.7194 1.50182.7185 1.50009.7174 1.00424.7177 1.50013.7180 1.50013.7175 1.00423.7185 1.50183.7131 1.50183.7174 1.50183.7180 1.50183.7194 1.51196.7131 1.50182.7177 1.51196.7174 1.51196.7175 1.51196.7177 1.51196.7180 1.51196.7194 1.51196.7220 1.00424.7174 1.00424.7180 1.50182.7194 1.50012.7185 1.50012.7194 1.50013.7194 1.50012.7175 1.50012.7177 1.50012.7180 1.50012.7220 1.50013.7174 1.50013.7177 1.50013.7220 1.50182.7184 1.00424.7185 1.50182.7175 1.50182.7180 1.50182.7220 1.50012.7174 1.50012.7184
1.00423.7130EA 1120 EUR
1.00423.7130 1.00424.7130 1.50009.7127 1.50009.7130 1.50012.7130 1.50013.7127 1.50013.7130 1.50013.7184 1.50182.7127 1.50182.7130 1.50183.7127 1.50183.7130 1.50009.7184 1.50183.7185 1.50182.7174 1.00423.7184 1.50009.7220 1.00424.7184 1.50183.7177 1.00424.7175 1.00424.7194 1.50182.7185 1.50009.7174 1.00424.7177 1.50013.7180 1.50013.7175 1.00423.7185 1.50183.7131 1.50183.7174 1.50183.7180 1.50183.7194 1.51196.7131 1.50182.7177 1.51196.7174 1.51196.7175 1.51196.7177 1.51196.7180 1.51196.7194 1.51196.7220 1.00424.7174 1.00424.7180 1.50182.7194 1.50012.7185 1.50012.7194 1.50013.7194 1.50012.7175 1.50012.7177 1.50012.7180 1.50012.7220 1.50013.7174 1.50013.7177 1.50013.7220 1.50182.7184 1.00424.7185 1.50182.7175 1.50182.7180 1.50182.7220 1.50012.7174 1.50012.7184
HPLC-kolonni, Hibar®, with Purospher® Sorbents
Kromatografiakolonnit
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of Purospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number MERC1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.