Phosphorus CRM, standard solutions, Supelco®
Toimittaja: Merck
1.25046.0100 1.25047.0100 1.25048.0100 1.25049.0100
1.25046.0100EA 55.4 EUR
1.25046.0100 1.25047.0100 1.25048.0100 1.25049.0100
Phosphorus CRM, standard solutions, Supelco®
Standardiaineet ja vertailumateriaalit Anionic Standards Phosphate Standards
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Sertifikaatit: Traceable to SRM from NIST and USP.